Skip to Content
Merck
All Photos(1)

Key Documents

8.45127

Sigma-Aldrich

Chlorosulfuric acid

≥98% (acidimetric)

Synonym(s):

Chlorosulfonic acid, Chlorosulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
116.53
MDL number:
UNSPSC Code:
12352100
EC Index Number:
232-234-6
Pricing and availability is not currently available.

Product Name

Chlorosulfonic acid, Msynthplus

grade

reagent

vapor pressure

3.08 hPa ( 20 °C)

Assay

≥98% (acidimetric)

form

liquid

potency

50 mg/kg LD50, oral (Rat)

pH

<1 (20 °C in H2O, Aqueous solution; rigorous decomposition)

bp

152 °C/1013 hPa (decomposition)

mp

-80 °C

density

1.75 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/ClHO3S/c1-5(2,3)4/h(H,2,3,4)

InChI key

XTHPWXDJESJLNJ-UHFFFAOYSA-N

General description

Msynthplus

Application

Chlorosulfonic acid Msynthplus. CAS 7790-94-5, pH < - 1 (H₂O, 20 °C) Aqueous solution,(rigorous decomposition).

Analysis Note

Colour (visual)colourlessAppearance of substance (visual)clearAssay (acidimetric)≥ 98.0 %Free SO₃≤ 1.0 %The analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service