Skip to Content
Merck
All Photos(1)

Key Documents

8.45019

Sigma-Aldrich

Aluminium chloride

(anhydrous, powder, ground) Msynthplus

Synonym(s):

Aluminium chloride, Hydrochloric acid aluminium salt anhydrous

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3
CAS Number:
Molecular Weight:
133.34
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-208-1
form:
powder
solubility:
450 g/L (decomposition)

vapor pressure

1 hPa ( 20 °C)

form

powder

potency

3450 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

pH

2.4 (20 °C, 100 g/L in H2O)

mp

180-181 °C (sublimed)

solubility

450 g/L (decomposition)

density

1.31 g/cm3 at 200 °C (liquid)

bulk density

1200 kg/m3

storage temp.

2-30°C

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Colour (visual): off white
Assay (complexometric): ≥ 98.0 %
Identity (ICP-OES): passes test
Pb (Lead): lot specific result
The analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service