Skip to Content
Merck
All Photos(1)

Key Documents

8.41538

Sigma-Aldrich

1-Benzothiophene

for synthesis

Synonym(s):

1-Benzothiophene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6S
CAS Number:
Molecular Weight:
134.20
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-395-7
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

autoignition temp.

510 °C

potency

1700 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

bp

222 °C

mp

28-32 °C

transition temp

flash point 91 °C

density

1.15 g/cm3

storage temp.

2-30°C

SMILES string

[s]1c2c(cc1)cccc2

InChI

1S/C8H6S/c1-2-4-8-7(3-1)5-6-9-8/h1-6H

InChI key

FCEHBMOGCRZNNI-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 29 °C
Melting range (upper value): ≤ 32 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service