Skip to Content
Merck
All Photos(1)

Key Documents

8.40157

Sigma-Aldrich

Dicyclopropylketone

for synthesis

Synonym(s):

Dicyclopropylketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10O
CAS Number:
Molecular Weight:
110.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
214-331-5
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

liquid

bp

160-162 °C/1013 hPa

transition temp

flash point 39 °C

density

0.968 g/cm3

storage temp.

2-30°C

SMILES string

O=C(C2CC2)C1CC1

InChI

1S/C7H10O/c8-7(5-1-2-5)6-3-4-6/h5-6H,1-4H2

InChI key

BIPUHAHGLJKIPK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.965 - 0.970
Identity (IR): passes test

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service