8.24509
(-)-trans-2-tert-Butylcyclohexanol
≥99% (GC)
Synonym(s):
(-)-trans-2-tert-Butylcyclohexanol
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
Assay
≥99% (GC)
form
solid
storage temp.
2-30°C
InChI
1S/C10H20O/c1-10(2,3)8-6-4-5-7-9(8)11/h8-9,11H,4-7H2,1-3H3/t8-,9-/m1/s1
InChI key
DLTWBMHADAJAAZ-RKDXNWHRSA-N
General description
for synthesis
Application
(-)-trans-2-tert-Butylcyclohexanol for synthesis. CAS 98104-30-4, molar mass 156.27 g/mol.
Analysis Note
Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Spec. rotation [α²0/D (c=1 in chloroform): -44.0 - -40.0 °
Identity (IR): passes test
Melting range (lower value): ≥ 54 °C
Melting range (upper value): ≤ 57 °C
Spec. rotation [α²0/D (c=1 in chloroform): -44.0 - -40.0 °
Identity (IR): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service