Skip to Content
Merck
All Photos(2)

Key Documents

8.21276

Sigma-Aldrich

3-Ethoxy-4-hydroxybenzaldehyde

for synthesis

Synonym(s):

3-Ethoxy-4-hydroxybenzaldehyde, Ethylvanillin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3-(C2H5O)-4-(OH)C6H3CHO
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352114
EC Index Number:
204-464-7

Assay

≥99.0% (GC)

form

crystals

potency

>3160 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

mp

75-77 °C

transition temp

flash point 145 °C

solubility

2.82 g/L

storage temp.

2-30°C

InChI

1S/C9H10O3/c1-2-12-9-5-7(6-10)3-4-8(9)11/h3-6,11H,2H2,1H3

InChI key

CBOQJANXLMLOSS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 75 °C
Melting range (upper value): ≤ 77 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service