Skip to Content
Merck
All Photos(4)

Documents

8.20957

Supelco

n-Pentane

EMPLURA®

Synonym(s):

Pentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
MDL number:
UNSPSC Code:
12352001
EC Index Number:
203-692-4
NACRES:
NA.21

vapor density

2.48 (vs air)

Quality Level

product line

EMPLURA®

Assay

≥99% (GC)

form

liquid

autoignition temp.

500 °F

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

~8.3 %

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

transition temp

flash point -48 °C (refers to pure substance)

solubility

0.4 g/L

density

0.626 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • The effect of hydrogen bonding on the diffusion of water in n-alkanes and n-alcohols measured with a novel single microdroplet method.: This research investigates the diffusion behavior of water in n-pentane and other n-alkanes, emphasizing the role of hydrogen bonding. The novel single microdroplet method used in this study provides detailed insights into the molecular interactions and diffusion processes (Su et al., 2010).

  • Enthalpy of solution of 1,4-naphthoquinone in CO2 + n-pentane in the critical region of the binary mixture: mechanism of solubility enhancement.: This paper explores the solubility mechanisms of 1,4-naphthoquinone in a CO2 and n-pentane mixture, particularly in the critical region. The study reveals how n-pentane enhances the solubility of organic compounds in supercritical CO2, with implications for industrial applications (Mu et al., 2004).

  • Metabolism of n-pentane by ethanol-inducible cytochrome P-450 in liver microsomes and reconstituted membranes.: This research investigates the metabolic pathways of n-pentane in the presence of ethanol-inducible cytochrome P-450. The findings elucidate the role of specific enzymes in the biotransformation of n-pentane, with significant implications for toxicology and pharmacology (Terelius & Ingelman-Sundberg, 1986).


Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): conforms
Density (d 20 °C/ 4 °C): 0.625 - 0.627

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service