Skip to Content
Merck
All Photos(2)

Documents

8.20597

Sigma-Aldrich

Ethyl 3,4,5-trihydroxybenzoate

for synthesis

Synonym(s):

Ethyl gallate, Gallic acid ethyl ester, 3,4,5-Trihydroxybenzoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3,4,5-(OH)3C6H2COOC2H5
CAS Number:
Molecular Weight:
198.17
Beilstein:
2116014
MDL number:
UNSPSC Code:
12352108
EC Index Number:
212-608-5
NACRES:
NA.22

Quality Level

Assay

≥98.0% (acidimetric)

form

powder

mp

150-152 °C

storage temp.

2-30°C

SMILES string

CCOC(=O)c1cc(O)c(O)c(O)c1

InChI

1S/C9H10O5/c1-2-14-9(13)5-3-6(10)8(12)7(11)4-5/h3-4,10-12H,2H2,1H3

InChI key

VFPFQHQNJCMNBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 150 °C
Melting range (upper value): ≤ 152 °C
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service