Skip to Content
Merck
All Photos(1)

Documents

8.20031

Sigma-Aldrich

Ethylenediammonium sulfate

for synthesis

Synonym(s):

Ethylenediammonium sulfate, 1.2-Diaminoethane sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H10N2O4S
CAS Number:
Molecular Weight:
158.18
UNSPSC Code:
12352107
EC Index Number:
244-733-6
NACRES:
NA.22

Quality Level

form

crystals

mp

225-228 °C

storage temp.

2-30°C

InChI

1S/C2H8N2.H2O4S/c3-1-2-4;1-5(2,3)4/h1-4H2;(H2,1,2,3,4)

InChI key

BNZCDZDLTIHJAC-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric after ion exchange): ≥ 97.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service