Skip to Content
Merck
All Photos(1)

Key Documents

8.18731

Sigma-Aldrich

Salicylic acid

for synthesis

Synonym(s):

Salicylic acid, 2-Hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6O3
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-712-3
NACRES:
NA.22
pH:
2.4 ( in H2O, saturated solution)
Pricing and availability is not currently available.

vapor pressure

<1 hPa ( 100 °C)

Quality Level

Assay

≥99% (acidimetric)

form

solid

autoignition temp.

500 °C

potency

1250-1580 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

technique(s)

tissue culture: suitable

pH

2.4 ( in H2O, saturated solution)

bp

211 °C/1013 hPa

mp

157-159 °C

transition temp

flash point 157 °C

solubility

water: 2 g/L at 20 °C

density

1.443 g/cm3 at 20 °C

bulk density

400‑500 kg/m3

storage temp.

2-30°C

SMILES string

[O-]c1c(cccc1)C(=O)O.[H+]

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Application


  • Salicylic Acid and Water Stress: Investigates the impact of salicylic acid on the morphophysiology and essential oil profile of Eryngium foetidum under water stress conditions, highlighting its role in enhancing plant resilience and secondary metabolite production (Santos et al., 2024).

  • Salicylic Acid in Plant Defense: Focuses on the usage of salicylic acid as an elicitor to boost the production of withanolides and phenolic acids in Withania somnifera, demonstrating its efficacy in improving secondary metabolite yield in plant biotechnological applications (Halder and Ghosh, 2024).

  • Salicylic Acid in Natural Deep Eutectic Solvents: Describes the application of natural deep eutectic solvents containing salicylic acid for forming aqueous biphasic systems, presenting a novel method for drug extraction and highlighting its potential in pharmaceutical manufacturing (Ghaffari et al., 2024).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service