Skip to Content
Merck
All Photos(1)

Key Documents

8.18700

Supelco

n-Amyl acetate

EMPLURA®

Synonym(s):

n-Amyl acetate, Pear oil, n-Pentyl acetate, Acetic acid pentyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COO(CH2)4CH3
CAS Number:
Molecular Weight:
130.18
MDL number:
UNSPSC Code:
12352108
EC Index Number:
211-047-3
NACRES:
NB.21
Assay:
≥98.0% (GC)
bp:
149 °C/1013 hPa
vapor pressure:
6 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

6 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

350 °C

potency

>1600 mg/kg LD50, oral (Rat)

expl. lim.

1.1-7.5 % (v/v)

bp

149 °C/1013 hPa

mp

-71 °C

transition temp

flash point 41 °C

solubility

10 g/L

density

0.87 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCCCC)C(=O)C

InChI

1S/C7H14O2/c1-3-4-5-6-9-7(2)8/h3-6H2,1-2H3

InChI key

PGMYKACGEOXYJE-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Application


  • Use of a habituation-dishabituation test to determine canine olfactory sensitivity.: This study by Moser et al. examines the use of a habituation-dishabituation test to measure olfactory sensitivity in dogs. The research explores the detection threshold of various odorants, including n-Amyl acetate, highlighting its application in behavioral studies involving canines (Moser et al., 2022).

  • Enhancement of olfaction by femtomolar concentrations of zinc ions.: Ishimaru investigates the effects of zinc ions on olfactory sensitivity, showing significant enhancement even at femtomolar concentrations. The study uses n-Amyl acetate as a reference odorant to demonstrate improved olfactory function (Ishimaru, 2022).

  • Generalization and discrimination tasks yield concordant measures of perceived distance between odours and their binary mixtures in larval Drosophila.: Chen and Gerber study the perception of odor mixtures in larval Drosophila, using n-Amyl acetate to assess generalization and discrimination tasks, which reflect the perceived distance between odors (Chen & Gerber, 2014).

  • SML resist processing for high-aspect-ratio and high-sensitivity electron beam lithography.: Mohammad et al. present advancements in SML resist processing, using n-Amyl acetate in the fabrication process for high-aspect-ratio and high-sensitivity applications in electron beam lithography (Mohammad et al., 2013).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.875 - 0.877
Identity (IR): passes test

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

105.8 °F

Flash Point(C)

41 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service