Skip to Content
Merck
All Photos(1)

Key Documents

8.14893

Sigma-Aldrich

α-Phellandrene

for synthesis

Synonym(s):

α-Phellandrene, 5-Isopropyl-2-methyl-1,3-cyclohexadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
MDL number:
UNSPSC Code:
12352002
EC Index Number:
202-792-5
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

form

liquid

potency

5700 mg/kg LD50, oral (Rat)

concentration

≥60.0% (GC)

bp

175 °C/1013 hPa

transition temp

flash point 47 °C

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[C@@H]1(CC=C(C=C1)C)C(C)C

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4-6,8,10H,7H2,1-3H3/t10-/m1/s1

InChI key

OGLDWXZKYODSOB-SNVBAGLBSA-N

Analysis Note

Assay (GC, area%): ≥ 60.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.835 - 0.850

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service