Skip to Content
Merck
All Photos(1)

Key Documents

8.14099

Sigma-Aldrich

2-Amino-2-methyl-1-propanol hydrochloride

for synthesis

Synonym(s):

2-Amino-2-methyl-1-propanol hydrochloride, AMP hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(NH2)CH2OH · HCl
CAS Number:
Molecular Weight:
125.60
MDL number:
UNSPSC Code:
12352107
EC Index Number:
221-713-5
NACRES:
NA.22
form:
powder
Pricing and availability is not currently available.

Quality Level

form

powder

mp

204 °C (External MSDS)

storage temp.

2-30°C

SMILES string

[Cl-].[N+H3]C(CO)(C)C

InChI

1S/C4H11NO.ClH/c1-4(2,5)3-6;/h6H,3,5H2,1-2H3;1H

InChI key

PMZIUAOBHNJYQT-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service