Skip to Content
Merck
All Photos(1)

Key Documents

8.08649

Sigma-Aldrich

Trioctylamine

for synthesis

Synonym(s):

Trioctylamine, N,N-Dioctyl-1-octanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H51N
CAS Number:
Molecular Weight:
353.67
MDL number:
UNSPSC Code:
12352116
EC Index Number:
214-242-1
NACRES:
NA.22
form:
liquid
Assay:
≥92.5% (GC)
Pricing and availability is not currently available.

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥92.5% (GC)

form

liquid

autoignition temp.

315 °C

mp

-39 °C

transition temp

flash point 168 °C

solubility

<0.0001 g/L

density

0.81 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCCCCCCC)(CCCCCCCC)CCCCCCCC

InChI

1S/C24H51N/c1-4-7-10-13-16-19-22-25(23-20-17-14-11-8-5-2)24-21-18-15-12-9-6-3/h4-24H2,1-3H3

InChI key

XTAZYLNFDRKIHJ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 92.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.808 - 0.813
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service