Skip to Content
Merck
All Photos(1)

Key Documents

8.07476

Sigma-Aldrich

Isopropylamine

for synthesis

Synonym(s):

Isopropylamine, 2-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
MDL number:
UNSPSC Code:
12352116
EC Index Number:
200-860-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

631 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

330 °C

potency

550 mg/kg LD50, oral (Rat)
>400 mg/kg LD50, skin (Rabbit)

expl. lim.

2-10.4 % (v/v)

pH

13 (20 °C, 700 g/L in H2O)

kinematic viscosity

0.47 cSt(20 °C)

bp

32 °C/1013 hPa

transition temp

flash point -37 °C

density

0.69 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

NC(C)C

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3

InChI key

JJWLVOIRVHMVIS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.687 - 0.688
Water (K. F.): ≤ 0.50 %
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-13.0 °F - closed cup

Flash Point(C)

<= -25 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service