Skip to Content
Merck
All Photos(1)

Documents

8.06356

Sigma-Aldrich

Sodium hydrogen sulfite

(39% solution in water) for synthesis

Synonym(s):

Sodium bisulfite, Sodium bisulfite solution, Bisulfite

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352302
NACRES:
NA.22

vapor pressure

40 hPa ( 20 °C)

Quality Level

form

liquid

pH

4.5 (20 °C in H2O)

mp

-44 °C

density

1.34 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • Pharmacokinetic studies: An LC-MS/MS method incorporating microelution-solid phase extraction was developed for the simultaneous plasma quantification of aripiprazole and its metabolite, dehydro-aripiprazole, indicating its application in human pharmacokinetic studies utilizing deuterated aripiprazole, such as Aripiprazole-d₈ (Wojnicz et al., 2018).

Analysis Note

Assay (iodometric): 37.0 - 42.0 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Supplementary Hazards

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service