Skip to Content
Merck
All Photos(1)

Key Documents

8.03225

Sigma-Aldrich

1,3-Dichlorobenzene

for synthesis

Synonym(s):

1,3-Dichlorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$175.95
500 MG
$250.70
1 G
$351.90
5 G
$1,256.95
10 G
$2,136.70

Vendor SKU: 45534-250MG

$175.95


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$175.95
500 MG
$250.70
1 G
$351.90
5 G
$1,256.95
10 G
$2,136.70

About This Item

Linear Formula:
1,3-(Cl)2C6H4
CAS Number:
Molecular Weight:
147.00
MDL number:
UNSPSC Code:
12352101
EC Index Number:
208-792-1

Vendor SKU: 45534-250MG

$175.95


Ships in 7 days - from Aldrich Partner.

vapor pressure

3 hPa ( 20 °C)

Assay

≥98.0% (GC)

autoignition temp.

>500 °C

potency

580 mg/kg LD50, oral (Rat)

bp

173 °C/1013 hPa

mp

-25 °C

transition temp

flash point 65 °C

density

1.29 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H

Application

1,3-Dichlorobenzene for synthesis. CAS 541-73-1, chemical formula 1,3-(Cl)₂C₆H₄.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.287 - 1.289
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Select a Product Number

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service