Skip to Content
Merck
All Photos(1)

Key Documents

8.02622

Sigma-Aldrich

1-Bromo-3-chloro-2-methylpropane

for synthesis

Synonym(s):

1-Bromo-3-chloro-2-methylpropane, 1-Chloro-3-bromo-2-methylpropane, 1-Chloro-2-methyl-3-bromopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH(CH3)CH2Br
CAS Number:
Molecular Weight:
171.46
UNSPSC Code:
12352101
EC Index Number:
230-224-6

Quality Level

Assay

≥98.0% (GC)

form

liquid

bp

157-159 °C/1013 hPa

transition temp

flash point >110 °C

density

1.48 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H8BrCl/c1-4(2-5)3-6/h4H,2-3H2,1H3

InChI key

ZKDOQFPDSUOLGF-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.481 - 1.484
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service