Skip to Content
Merck
All Photos(1)

Documents

8.01976

Sigma-Aldrich

Isobutyryl chloride

for synthesis

Synonym(s):

Isobutyryl chloride, 2-Methylpropionyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOCl
CAS Number:
Molecular Weight:
106.55
MDL number:
UNSPSC Code:
12352106
EC Index Number:
201-194-1

vapor pressure

57 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

325 °C

potency

>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.1-8.2 % (v/v)

pH

<7 ( in H2O)

mp

-90 °C

transition temp

flash point 8.5 °C

density

1.01 g/cm3 at 20 °C

InChI

1S/C4H7ClO/c1-3(2)4(5)6/h3H,1-2H3

InChI key

DGMOBVGABMBZSB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.008 - 1.009
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service