Skip to Content
Merck
All Photos(1)

Documents

8.00465

Sigma-Aldrich

Anthraquinone

for synthesis

Synonym(s):

Anthraquinone, 9,10-Dihydro-9,10-anthracenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8O2
CAS Number:
Molecular Weight:
208.21
MDL number:
UNSPSC Code:
12352115
EC Index Number:
201-549-0
NACRES:
NA.22

vapor pressure

<1 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (HPLC)

form

powder

autoignition temp.

650 °C

potency

>5000 mg/kg LD50, oral (Rat)

transition temp

flash point 250 °C

solubility

0.00007 g/L

density

1.26 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

no temp limit

InChI

1S/C14H8O2/c15-13-9-5-1-2-6-10(9)14(16)12-8-4-3-7-11(12)13/h1-8H

InChI key

RZVHIXYEVGDQDX-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

482.0 °F - closed cup

Flash Point(C)

250 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service