Skip to Content
Merck
All Photos(1)

Key Documents

73295BC

Millipore

mEHEC® 47.4 g Media Scoop

BioControl®, For use with mEHEC®

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
R$722.00

R$722.00


Estimated to ship onMay 06, 2025


Request more information

Select a Size

Change View
1 EACH
R$722.00

About This Item

UNSPSC Code:
41122419
NACRES:
NA.74

R$722.00


Estimated to ship onMay 06, 2025


Request more information

Quality Level

storage condition

room temperature

application(s)

food and beverages
pathogen testing

compatibility

BioControl, For use with mEHEC

Components

One 47.4 g scoop

Compatibility

BioControl® GDS

Testing Method

PCR

Legal Information

BioControl is a registered trademark of BioControl Systems, Inc.
mEHEC is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service