Skip to Content
Merck
All Photos(1)

Documents

71574

Sigma-Aldrich

pIEx-10 Ek/LIC Vector Kit - Novagen®

The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera derived insect cells.

Synonym(s):

pIEx vectors, pIEx-10 Ek/LIC vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.54

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze

shipped in

dry ice

storage temp.

−70°C

General description

The pIEx-10 Ek/LIC vector is an Ek/LIC version of pIEx-10. The pIEx vectors are designed for cloning and high-level expression of proteins in transiently transfected Spodoptera-derived insect cells. Transcription is driven by the AcNPV-derived hr5 enhancer and immediate early promoter, IE1. The pIEx-10 Ek/LIC vector contains the coding sequence for the mouse IgM signal sequence to allow secretion of the expressed protein. It also encodes an N-terminal 8 aa Strep•Tag II coding sequence followed by an enterokinase protease cleavage site and a multiple cloning site, with the option to include a C-terminal His•Tag® coding sequence with 10 histidines.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

HIS TAG is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product contains genetically modified organisms (GMO). Within the EU GMOs are regulated by Directives 2001/18/EC and 2009/41/EC of the European Parliament and of the Council and their national implementation in the member States respectively. This legislation obliges MilliporeSigma to request certain information about you and the establishment where the GMOs are being handled. Clickhere for Enduser Declaration (EUD) Form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service