Skip to Content
Merck
All Photos(1)

Key Documents

70594

Sigma-Aldrich

Introductory pSTBlue-1 AccepTor Vector Kit - Novagen

Synonym(s):

AccepTor Vector Cloning Kits, AccepTor Vector Kits, pSTBlue-1 Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.54
Pricing and availability is not currently available.

manufacturer/tradename

Novagen®

Quality Level

storage condition

OK to freeze

shipped in

dry ice

storage temp.

−70°C

General description

The AccepTor Vector Cloning Kits are designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases (i.e. KOD XL polymerase and native and recombinant Taq polymerases), which leave single 3′-dA overhangs on the reaction products. The AccepTor Vectors enable direct ligation by virtue of single 3′-dU overhangs that anneal efficiently with 3′-dA extensions on PCR products. The dU residues are converted to dT residues in vivo following transformation.

AccepTor Vectors are supplied in a ready-to-ligate format. Simply mix the vector with your PCR product, add Clonables 2X Ligation Premix (Cat No. 70573), and transform into NovaBlue Singles Competent Cells (Cat. No. 70181). The entire procedure, from PCR product to plating transformants, can be performed in as little as 40 minutes with minimal pipetting.

AccepTor Vector Kits are available in an introductory 10-reaction size as well as 20- and 40-reaction configurations. The AccepTor Vectors are also available separately without the ligation and transformation components. Also see pSTBlue-1 AccepTor Vector Giga Kit (Cat. No. 71228) for information on using higher-efficiency competent cells.

Two different vectors, pSTBlue-1 and pETBlue-1, are available as AccepTor Vector kits. Each is carefully prepared and tested for optimal cloning efficiency and provides easy visualization of recombinants by blue/white screening using lacZ α-complementation. The pSTBlue-1 is a general purpose vector with dual opposed T7 and SP6 promoters, both with amp and kan resistance, and an array of flanking restriction sites. The pETBlue-1 vector is a novel plasmid specifically designed to enable high-level T7 RNA polymerase-driven expression of target genes, while providing the convenience of a blue/white screening method. Screening is independent of expression because the associated T7lac expression promoter is in an opposed orientation relative to the E. coli promoter. pETBlue-1 facilitates the expression of native unfused proteins and allows for convenient subcloning of target genes already fused to existing detection and purification tags. An EcoR V cloning site is appropriately spaced downstream of an E. coli ribosome binding site. Inserts must encode an ATG start codon at their 5′ end if expression is desired. The sequence is numbered from the first base of the T7 promoter sequence.

Features and Benefits

  • No restriction enzymes or special primers
  • Direct ligation of PCR product with vector
  • Compatible with polymerases that leave single 3´-dA overhangs
  • Blue/white screening with pSTBlue-1 or pETBlue-1 vectors
  • Simple protocol takes as little as 40 minutes from PCR product to plating transformants

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Legal Information

This product is covered by U.S. Patent 5,856,144 owned by EMD Chemicals Inc. or its Affiliates.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service