Skip to Content
Merck
All Photos(1)

Key Documents

691550-M

Sigma-Aldrich

Zinc (II) Protoporphyrin IX

A potent and selective inhibitor of heme oxygenase, the enzyme which generates carbon monoxide (CO) and biliverdin.

Synonym(s):

Zinc (II) Protoporphyrin IX, ZnPP-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H32N4O4Zn
CAS Number:
Molecular Weight:
626.03
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark red

solubility

DMF: soluble
pyridine: soluble

storage temp.

−20°C

SMILES string

[Zn+2].[n-]1c2cc3nc(cc4[n-]c(cc5nc(cc1c(c2CCC(=O)O)C)c(c5C=C)C)c(c4C=C)C)c(c3CCC(=O)O)C

InChI

1S/C34H34N4O4.Zn/c1-7-21-17(3)25-13-26-19(5)23(9-11-33(39)40)31(37-26)16-32-24(10-12-34(41)42)20(6)28(38-32)15-30-22(8-2)18(4)27(36-30)14-29(21)35-25;/h7-8,13-16H,1-2,9-12H2,3-6H3,(H4,35,36,37,38,39,40,41,42);/q;+2/p-2/b25-13?,26-13-,27-14?,28-15-,29-14-,30-15?,31-16-,32-16?;

InChI key

FUTVBRXUIKZACV-RGUWPKJZSA-L

General description

A potent and selective inhibitor of heme oxygenase, the enzyme which generates carbon monoxide (CO) and biliverdin. Inhibits soluble guanylyl cyclase and produces a time- and concentration-dependent inactivation of all three isoforms of nitric oxide synthase (IC50 = 800 nM, 4 µM, and 5 µM for nNOS, iNOS, and eNOS, respectively). Inhibits relaxation induced by acetylcholine, ANP, and VIP in isolated rat aorta. Blocks interleukin-1 and prevents induction of long-term potentiation. Also useful as an indicator of iron-deficient erythropoiesis. Does not cross the blood-brain barrier.
A potent and selective inhibitor of heme oxygenase, the enzyme which generates carbon monoxide (CO) and biliverdin. Inhibits soluble guanylyl cyclase. Produces a time- and concentration-dependent inactivation of all three isoforms of nitric oxide synthase (IC50 = 0.8 µM, 4.0 µM, and 5.0 µM for nNOS, iNOS, and eNOS, respectively). Inhibits relaxation induced by acetylcholine, ANP, and VIP in isolated rat aorta. Blocks interleukin-1 and prevents induction of long-term potentiation. Also useful as an indicator of iron deficient erythropoiesis. Does not cross the blood-brain barrier.

Biochem/physiol Actions

Primary Target
heme oxygenase
Target IC50: 800 nM, 4 µM, and 5 µM for nNOS, iNOS, and eNOS, respectively

Warning

Toxicity: Standard Handling (A)

Other Notes

Wolff, D.J., et al. 1996. Arch. Biochem. Biophys.333, 27.
Bing, O., et al. 1995. NeuroReport 6, 1369.
Hinchliffe, R.F., et al. 1995. Pediatr. Hematol. Oncol. 12, 455.
Kadoya, C., et al. 1995. Stroke 26, 1035.
Ny, L., et al. 1995. Br. J. Pharmacol. 115, 186.
Luo, D., and Vincent, S.R. 1994. Eur. J. Pharmacol. 267, 263.
Marks, G.S., et al. 1993. Can. J. Physiol. Pharmacol. 71, 753.
Zhuo, M., et al. 1993. Science 260, 1946.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 121°C 15분 고압멸균 오토클레이브를 돌리려고하는데 멸균해도 화학구조상 문제가 없을지에 대하여 여쭙습니다.

    1 answer
    1. This material has not been evaluated for stability under high heat conditions. Stock solutions may be filter sterilized.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service