Skip to Content
Merck
All Photos(1)

Key Documents

676472

Sigma-Aldrich

VEGF₁₆₅, Human, Recombinant, S. frugiperda

Synonym(s):

Vascular Endothelial Growth Factor₁₆₅, rhVEGF, rhVPF, rh-Vasculotropin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
Pricing and availability is not currently available.

Assay

≥97% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤1 EU/μg Endotoxin (EU/μg VEGF)

shipped in

ambient

storage temp.

−70°C

General description

Recombinant, human VEGF165 (165 amino acid splice variant of VEGF) expressed in S. frugiperda insect cells. A heparin-binding glycoprotein with potent angiogenic, endothelial cell-specific mitogenic, and vascular permeability-enhancing activities. Increases endothelial cell proliferation after vascular injury. Supplied as a glycosylated, disulfide-linked homodimer.

Biochem/physiol Actions

ED₅₀ = 2.0-6.0 ng/ml as measured by its ability to stimulate ³H-thymidine incorporation into human umbilical vein endothelial cells

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered 30% acetonitrile, 0.1% TFA, 50 µg BSA/µg VEGF.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at -70°C.
Reconstitute to a final concentration of ≥1 µg/ml with sterile PBS containing ≥0.1% HSA or BSA.

Other Notes

Liu, X., et al. 2001. J. Urology166, 354.
Burke, P.A., et al. 1995. Biochem. Biophys. Res. Commun. 207, 348.
Neufeld, G., et al. 1994. Prog. Growth Factor Res.5, 89.
Leung, D.W., et al. 1989. Science 246, 1306.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service