Skip to Content
Merck
All Photos(1)

Key Documents

65347-M

Sigma-Aldrich

Alcohol 100%

HARLECO® Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols

Synonym(s):

Alcohol, Dehydration

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
41116121
application(s):
diagnostic assay manufacturing
hematology
histology
Pricing and availability is not currently available.

Quality Level

form

liquid

shelf life

18 mo. (from manufacture date)

composition

ethanol, 89.0-92.0%
isopropyl alcohol, 4.0-6.0%
methanol, 3.5-5.5%

color

APHA: ≤10

transition temp

flash point 12 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

10-30°C

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

HARLECO® Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols
Part of the QSM Histology stain line. Used to hydrate or dehydrate tissue samples in tissue processing and staining protocols.

Analysis Note

Expiration date18 months from mfg dateAssay (Ethanol)89.0-92.0%Assay (Isopropyl alcohol)4.0-6.0%Assay (Methanol)3.5-5.5%Color (APHA)10APHA maxFormClear liquidInfrared SpectrumConforms to standardResidue after evaporation0.001% maxWater (H2O)0.075% max

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service