Skip to Content
Merck
All Photos(1)

Documents

65044-M

Sigma-Aldrich

Hemacolor® Stain Set

Synonym(s):

Hematology Stain, Hematology Solutions

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Application

HARLECO® Complete stain set for quick differential staining of peripheral blood cells
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Complete stain set for quick differential staining of peripheral blood cells. Similar results to that of Wright/Wright-Giemsa Stain.

Components

Methanol fixative, eosin stain and methylene blue stain.

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany
HEMACOLOR is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service