Skip to Content
Merck
All Photos(1)

Key Documents

6.18550

Sigma-Aldrich

Triethylamine

for chromatography

Synonym(s):

Triethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)3N
CAS Number:
Molecular Weight:
101.19
MDL number:
UNSPSC Code:
12352116
Pricing and availability is not currently available.

grade

for chromatography

Quality Level

vapor density

3.48 (vs air)

vapor pressure

72 hPa ( 20 °C)

form

liquid

expl. lim.

1.2-9.3 % (v/v)

technique(s)

HPLC: suitable

pH

(12.7 at 100 g/l at 15 °C (59 °F))

bp

89.3 °C (192.7 °F)

solubility

water: soluble (112.4 g/l at 20 °C (68 °F))

density

0.72 g/cm3 (at 25 °C (77 °F))

application(s)

general analytical

SMILES string

N(CC)(CC)CC

InChI

1S/C6H15N/c1-4-7(5-2)6-3/h4-6H2,1-3H3

InChI key

ZMANZCXQSJIPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylamineis used as a mobile phase reagent in reversed-phase liquid chromatography todeactivate free silanol groups present in the stationary phase. It can also beused as a micellar mobile phase.[1][2]

Application

Triethylamine can be used as:
  • An environmentally friendly mobile phase component in liquid chromatographic technique for separating analgesic and antipyretic drugs such as propyphenazone, caffeine, paracetamol, and its degradation impurity 4-aminophenol.[3]
  • Mobile phase component in the determination of polyamines in thermosetting polymers.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - closed cup

Flash Point(C)

-11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green chromatography separation of analytes of greatly differing properties using a polyethylene glycol stationary phase and a low-toxic water-based mobile phase
Satinsky D, et al.
Analytical and Bioanalytical Chemistry, 405, 6105-6115 (2013)
Improved efficiency in micellar liquid chromatography using triethylamine and 1-butanol as mobile phase additives to reduce surfactant adsorption
Thomasa DP and Foleya JP
Journal of Chromatography A, 1205, 36-45 (2008)
Liquid chromatography method to determine polyamines in thermosetting polymers
Dopico-Garcia MS, et al.
Analytica Chimica Acta, 667(1-2), 123-129 (2010)
System peaks observed in capillary liquid chromatography with eluents containing triethylamine
Kafkova, B, et al.
Chromatographia, 56, 445-447 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service