Skip to Content
Merck
All Photos(2)

Key Documents

613550

Sigma-Aldrich

TMEM16A Activator, Eact

The TMEM16A Activator, Eact modulates the biological activity of TMEM16A. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

TMEM16A Activator, Eact, ANO1 Activator, Eact, 3,4,5-trimethoxy-N-(2-methoxyethyl)-N-(4-phenylthiazol-2-yl)benzamide, TMEM16A Activator, Eact

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O5S
CAS Number:
Molecular Weight:
428.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)c3ccccc3)N(CCOC)C(=O)c2cc(c(c(c2)OC)OC)OC

InChI

1S/C22H24N2O5S/c1-26-11-10-24(22-23-17(14-30-22)15-8-6-5-7-9-15)21(25)16-12-18(27-2)20(29-4)19(13-16)28-3/h5-9,12-14H,10-11H2,1-4H3

InChI key

ZUXNHFFVQWADJL-UHFFFAOYSA-N

General description

An N-aroylaminothiazole that strongly activates TMEM16A (ANO1) calcium-activated chloride channel (CaCC) at 0 Ca2+ concentration (Ca2+ independent), with the capability of producing large and sustained CaCC Cl-1 currents (EC50 = 3 µM) in TMEM16A-expressing Fisher Rat Thyroid (FRT) Cells without elevation of cytoplasmic Ca2+ level, and in human A253 cells. Shown to increase CaCC conductance in human salivary and airway submucosal gland epithelial cells, IL-4 treated bronchial cells, and stimulated submucosal gland secretion in human bronchi and smooth muscle contraction in mouse intestine. Activation effect can be abolished by the addition of a TMEM16A inhibitor T16A-A01 at 10 µM in TMEM16A-transfected cells and in cultures of human salivary gland and IL-4-treated bronchial epithelia, but not in intestine. Does not affect CFTR Cl- conductance or ENaC Na+ conductance.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Namkung, W., et al. 2011. FASEB J.25, 4048.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service