Skip to Content
Merck
All Photos(1)

Documents

605157

Sigma-Aldrich

Thrombin, Bovine

Synonym(s):

Thrombin, Bovine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.25

form

lyophilized

Quality Level

specific activity

≥1800 NIH units/mg protein
≥50 NIH units/mg dry wt

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

0.05 M NaCl: ≤2000 NIH units/mL
water: >2000 NIH units/mL

shipped in

ambient

storage temp.

−20°C

General description

Native, bovine thrombin. Key enzyme in the coagulation cascade. Converts fibrinogen into fibrin and activates factor XIII, which cross-links and stabilizes the fibrin polymer. Has a pH of 6.0-7.5 in a 7% solution.
Native, bovine thrombin. Key enzyme that activates factor XIII and converts fibronogen into fibrin.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is determined by comparison with a standard curve prepared using the Bureau of Biologics standard thrombin.

Physical form

Lyophilized from 50 mM Sodium Citrate, 0.2 M NaCl, 0.1% PEG-8000, pH 6.5.

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 2 months at -70°C.





From Catalog:

Desc. Field- added "Key enzyme that activates factor XIII and converts fibronogen into fibrin."

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijia Luo et al.
Communications biology, 3(1), 114-114 (2020-03-12)
Exosomes are secreted extracellular vesicles with lipid bilayer membranes. They are emerging as a new category of messengers that facilitate cross-talk between cells, tissues, and organs. Thus, a critical demand arises for the development of a sensitive and non-invasive tracking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service