5985-OP
Magnesium Chloride Solution
OmniPur® Grade, Sterile solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352101
grade:
reagent grade
Recommended Products
grade
reagent grade
Quality Level
manufacturer/tradename
Calbiochem®
color
clear to haze-free
shipped in
ambient
General description
Sterile solution ideal for preparation of competent cells for transfection. Molecular biology grade.
Specifications:
DNase: None detected
Concentration (molarity): 0.98-1.10M
Protease: None detected
RNase: None detected
Sterility: To pass test
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Specifications:
DNase: None detected
Concentration (molarity): 0.98-1.10M
Protease: None detected
RNase: None detected
Sterility: To pass test
Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service