Skip to Content
Merck
All Photos(1)

Key Documents

581004

Sigma-Aldrich

Telomerase Inhibitor III, Sodium Salt

The Telomerase Inhibitor III, Sodium Salt controls the biological activity of Telomerase. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

TAG-6, 5ʹ-d(TTAGGG)-3ʹ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

solubility

water: soluble

shipped in

wet ice

storage temp.

−70°C

General description

A short, cell-permeable hexameric phosphorothioate oligonucleotide (PS-ODN) telomere mimic that inhibits telomerase activity in cell lysates and lengthens cell doubling time in vitro and in vivo at concentrations of less than 2.5 µM. Note: 150 nmol = 285 µg.
M.W. ~1900.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Telomerase activity in cell lysates
Product competes with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Sequence

5′-d(TTAGGG)-3′

Other Notes

Mata, J.E., et al. 1997. Toxicol. Appl. Pharmacol. 144, 189.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manuel Bernabé-García et al.
Molecular oncology, 15(7), 1818-1834 (2021-03-14)
Telomerase reverse transcriptase (TERT) maintains telomere homeostasis, thus ensuring chromosome stability and cell proliferation. In addition, several telomere-independent functions of human TERT have been described. In this study, we report that TERT binds directly to the TCF binding elements located

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service