567706
Sphingomyelin, Bovine Spinal Cord
Contains primarily nervonic and stearic acids.
Synonym(s):
Sphingomyelin, Bovine Spinal Cord, N-Acyl-sphingosine-1-phosphorylcholine
About This Item
Recommended Products
Quality Level
Assay
≥98% (TLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
ethanol: 1 mg/mL
acetic acid: 5 mg/mL
chloroform: 5 mg/mL
shipped in
ambient
storage temp.
−20°C
SMILES string
[P](=O)(OC[C@H](NC=O)[C@H](O)\C=C\CCCCCCCCCCCCC)(OCC[N+](C)(C)C)O
InChI
1S/C24H49N2O6P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(28)23(25-22-27)21-32-33(29,30)31-20-19-26(2,3)4/h17-18,22-24,28H,5-16,19-21H2,1-4H3,(H-,25,27,29,30)/p+1/b18-17+/t23-,24+/m0/s1
InChI key
LRYZPFWEZHSTHD-HEFFAWAOSA-O
General description
Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.
Biochem/physiol Actions
Precursor to ceramide second messengers via the action of sphingomyelinase.
Warning
Reconstitution
Other Notes
Naundorf, A. and Klaffke, W. 1996. Carbohydr. Res. 285, 141.
Slife, C.W., et al. 1989. J. Biol. Chem.264, 10371.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service