Skip to Content
Merck
All Photos(1)

Key Documents

567418-M

Millipore

Sodium Acetate, Anhydrous, Molecular Biology Grade

Commonly used in several applications, including precipitation of nucleic acids.

Synonym(s):

Sodium Acetate, Anhydrous, Molecular Biology Grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H3NaO2
CAS Number:
Molecular Weight:
82.03
MDL number:
UNSPSC Code:
12352302

grade

Molecular Biology

Quality Level

Assay

≥99% (perchloric acid titration)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: soluble

cation traces

heavy metals: ≤0.001%

absorption

≤0.01 at 260 nm in H2O at 100 mM
≤0.01 at 280 nm in H2O at 100 mM

foreign activity

DNases, proteases, RNases, none detected

storage temp.

10-30°C

SMILES string

[Na+].[O-]C(=O)C

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

General description

Commonly used in several applications including precipitation of nucleic acids. Absorbance (260 nm and 280 nm): ≤0.01
Commonly used in several applications, including precipitation of nucleic acids.

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service