Skip to Content
Merck
All Photos(1)

Key Documents

559387

Sigma-Aldrich

SB 202474 - CAS 172747-50-1 - Calbiochem

Synonym(s):

4-Ethyl-2(p-methoxyphenyl)-5-(4ʹ-pyridyl)-IH-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Assay

≥97% (HPLC)

Quality Level

form

solid

potency

34 nM IC50

color

off-white

solubility

DMSO: 25 mg/mL
methanol: soluble

General description

A negative control for MAP kinase inhibition studies.
A negative control for SB202190 and SB203580 in p38 MAP kinase inhibition studies.

Biochem/physiol Actions

p38β

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Nath et al.
Cellular & molecular biology letters, 6(2), 173-184 (2001-09-07)
The mitogen-activated protein kinase (MAPK) cascades are thought to be important mediators in the transduction of extracellular signals into cellular responses. The p38 kinase, a member of the MAPK superfamily, is activated by a wide variety of extracellular stimuli and
R Yu et al.
The Journal of biological chemistry, 275(4), 2322-2327 (2000-01-25)
Phase II drug-metabolizing enzymes, such as glutathione S-transferase and quinone reductase, play an important role in the detoxification of chemical carcinogens. The induction of these detoxifying enzymes by a variety of agents occurs at the transcriptional level and is regulated
J C Lee et al.
Nature, 372(6508), 739-746 (1994-12-22)
Production of interleukin-1 and tumour necrosis factor from stimulated human monocytes is inhibited by a new series of pyridinyl-imidazole compounds. Using radiolabelled and radio-photoaffinity-labelled chemical probes, the target of these compounds was identified as a pair of closely related mitogen-activated
Sarah Lockhead et al.
Cell reports, 32(2), 107901-107901 (2020-07-16)
Protein synthesis inhibitors (e.g., cycloheximide) block mitotic entry, suggesting that cell cycle progression requires protein synthesis until right before mitosis. However, cycloheximide is also known to activate p38 mitogen-activated protein kinase (MAPK), which can delay mitotic entry through a G2/M

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service