Skip to Content
Merck
All Photos(1)

Key Documents

540222

Sigma-Aldrich

Puromycin, Dihydrochloride

Puromycin, Dihydrochloride, CAS 58-58-2, is An aminonucleoside antibiotic that inhibits protein synthesis by blocking the translation step and causes premature release of nascent polypeptide chains.

Synonym(s):

Puromycin, Dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29N7O5 · 2HCl
CAS Number:
Molecular Weight:
544.43
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white to off-white

solubility

methanol: 20 mg/mL
water: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

InChI

1S/C22H29N7O5.2ClH/c1-28(2)19-17-20(25-10-24-19)29(11-26-17)22-18(31)16(15(9-30)34-22)27-21(32)14(23)8-12-4-6-13(33-3)7-5-12;;/h4-7,10-11,14-16,18,22,30-31H,8-9,23H2,1-3H3,(H,27,32);2*1H/t14-,15+,16?,18+,22+;;/m0../s1

InChI key

MKSVFGKWZLUTTO-USYAMCSGSA-N

General description

Prokaryotic and eukaryotic protein synthesis inhibitor. Nucleoside that resembles the aminoacyl-adenylyl terminus of aminoacyl-tRNA and competes for binding to the "A site" of the large ribosomal subunit. Its incorporation into a growing polypeptide results in termination of chain elongation and release of the nascent polypeptidyl-puromycin. Used for selection of mammalian cell lines expressing a transfected pac gene (Streptomyces alboniger), whose product, puromycin acetyltransferase, inactivates the drug via acetylation. Induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells. Like cycloheximide, puromycin is also used as a general protein synthesis inhibitor in numerous cellular assays.
Protein synthesis inhibitor. Inhibits translation by causing premature release of nascent polypeptide chains. Induces DNA fragmentation in thymocytes and in human HL-60 leukemia cells.
Puromycin, Dihydrochloride, CAS 58-58-2, is An aminonucleoside antibiotic that inhibits protein synthesis by blocking the translation step and causes premature release of nascent polypeptide chains.

Biochem/physiol Actions

Primary Target
protein synthesis

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, sterilize by filtration through a 0.22 µm pore-size filter, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Chow, S.C., et al. 1995. Exp. Cell Res. 216, 149.
Constam, D.B., et al. 1995. J. Biol. Chem.270, 26931.
Wirth, M., et al. 1994. J. Virol.68, 566.
Claeyssens, S., et al. 1993. FEBS Lett.315, 7.
Hechler, U., et al. 1993. Biochem. Biophys. Res. Commun.194, 1305.
Kaufman, S.H., et al. 1993. Cancer Res.53, 3976.
de la Luna, S. and Ortín, J. 1992. Methods Enzymol.216, 376.
Kalpaxis, D.L. and Drainas, D. 1992. Arch. Biochem. Biophys.300, 629.
Shipston, M.J. and Antoni, F.A. 1992. Biochem. Biophys. Res. Commun.189, 1382.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Chen et al.
BMC medicine, 20(1), 68-68 (2022-02-17)
Genome-wide association studies (GWASs) have identified multiple risk loci for Parkinson's disease (PD). However, identifying the functional (or potential causal) variants in the reported risk loci and elucidating their roles in PD pathogenesis remain major challenges. To identify the potential
Rui Chen et al.
Genome medicine, 14(1), 53-53 (2022-05-20)
Genome-wide association studies (GWASs) have identified multiple risk loci for bipolar disorder (BD). However, pinpointing functional (or causal) variants in the reported risk loci and elucidating their regulatory mechanisms remain challenging. We first integrated chromatin immunoprecipitation sequencing (ChIP-Seq) data from
Ying Huang et al.
Frontiers in physiology, 12, 697139-697139 (2021-09-08)
Our previous study found that acupuncture with low frequency electrical stimulation (Acu/LFES) prevents muscle atrophy by attenuation of protein degradation in mice. The current study examines the impact of Acu/LFES on protein synthesis. C57/BL6 mice received Acu/LFES treatment on hindlimb
Hotaka Kobayashi et al.
Nature communications, 13(1), 1435-1435 (2022-03-19)
MicroRNAs (miRNAs) are small non-coding RNAs, which regulate the expression of thousands of genes; miRNAs silence gene expression from complementary mRNAs through translational repression and mRNA decay. For decades, the function of miRNAs has been studied primarily by ensemble methods
Ting Zhu et al.
Veterinary research, 52(1), 148-148 (2021-12-22)
Viruses have evolved multiple strategies to manipulate their host's translational machinery for the synthesis of viral proteins. A common viral target is the alpha subunit of eukaryotic initiation factor 2 (eIF2α). In this study, we show that global protein synthesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service