Skip to Content
Merck
All Photos(1)

Key Documents

5.38439

Sigma-Aldrich

RV1088

Synonym(s):

RV1088, N-[4-[[4-[[[[3-(1,1-dimethylethyl)-1-(4-methylphenyl)-1H-pyrazol-5-yl]amino]carbonyl]amino]-1-naphthalenyl]oxy]-2-pyridinyl]-2-methoxyacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H34N6O4
CAS Number:
Molecular Weight:
578.66
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pink-brown

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, naphthalene based compound that acts as a narrow spectrum kinase inhibitor and reduces pro-inflammatory cytokine synthesis in LPS-activated primary human macrophages (IC50 = 0.63 and 1.66 nM for TNFα and IL-6), monocytes (IC50 = 0.97, 1.1, and 1.32 nM for TNFα, IL-6, and IL-8), and synovial fibroblasts (5.45 and 1.61 nM for IL-6 and IL-8). Shown to simultaneously inhibit the activity of p38α, p38γ, and Src (c-Src and Hck, and Syk). Also reported to inhibit antigen-induced release of beta-Hex, LTC4, and PGD2 (IC50 = 18, 11, and 21 nM, respectively) in cord blood derived mast cells.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
p-38α, p38γ, c-Src, Hck and SyK
Reversible: yes
Target IC50: 5.45 nM, 1.61 nM for IL-6, IL-8 inhibition respectively in activated rheumatoid arthritis synovial fibroblasts

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

To, W. S., et al. 2015. Br. J. Pharmacol.172, 3805.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service