Skip to Content
Merck
All Photos(1)

Key Documents

537011

Sigma-Aldrich

Prolyl Endopeptidase Inhibitor II

The Prolyl Endopeptidase Inhibitor II, also referenced under CAS 108708-25-4, controls the biological activity of Prolyl Endopeptidase. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Prolyl Endopeptidase Inhibitor II, Z-PP-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22N2O4
CAS Number:
Molecular Weight:
330.38
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥97% (HPLC)

form

oil

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

colorless to off-white

solubility

DMSO: 10 mg/mL
DMF: soluble
ethyl acetate: soluble

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable dipeptide aldehyde that acts as a specific, potent, slow and tight-binding transition state analog inhibitor of prolyl endopeptidase (Ki = 350 pM and 500 pM for mouse brain and human brain prolyl endopeptidase, respectively). Reported to form a hemiacetal with the active-site serine.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Mouse brain prolylendooeotidase
Product does not compete with ATP.
Reversible: no
Target Ki: 350 pM and 500 pM for mouse brain and human brain prolyl endopeptidase, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Pro-Pro-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fülöp, V., et al. 1998. Cell94, 161.
Kahyaoglu, A., et al. 1997. Biochem. J.322, 839.
Bakker, A.V., et al. 1990. Biochem. J.271, 559.
Wilk, S., and Orlowski, M. 1983. J. Neurochem.41, 69.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zehavit Goldberg et al.
International journal of molecular sciences, 23(2) (2022-01-22)
The aim of this study was to characterize the distribution of the thrombin receptor, protease activated receptor 1 (PAR1), in the neuroretina. Neuroretina samples of wild-type C57BL/6J and PAR1-/- mice were processed for indirect immunofluorescence and Western blot analysis. Reverse
Valery Golderman et al.
Biomedicines, 10(6) (2022-06-25)
Thrombin is present in peripheral nerves and is involved in the pathogenesis of neuropathy. We evaluated thrombin activity in skin punch biopsies taken from the paws of male mice and rats and from the legs of patients with suspected small-fiber
Chenxi Zhao et al.
Journal of neuroinflammation, 19(1), 189-189 (2022-07-17)
Nafamostat mesylate (nafamostat, NM) is an FDA-approved serine protease inhibitor that exerts anti-neuroinflammation and neuroprotective effects following rat spinal cord injury (SCI). However, clinical translation of nafamostat has been limited by an unclear administration time window and mechanism of action. Time
Mouhannad Malek et al.
Nature communications, 12(1), 2673-2673 (2021-05-13)
Vesicular traffic and membrane contact sites between organelles enable the exchange of proteins, lipids, and metabolites. Recruitment of tethers to contact sites between the endoplasmic reticulum (ER) and the plasma membrane is often triggered by calcium. Here we reveal a
Chenxi Zhao et al.
Neural regeneration research, 19(2), 434-439 (2023-07-25)
Argatroban is a synthetic thrombin inhibitor approved by U.S. Food and Drug Administration for the treatment of thrombosis. However, whether it plays a role in the repair of spinal cord injury is unknown. In this study, we established a rat

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service