Skip to Content
Merck
All Photos(1)

Key Documents

516724-M

Millipore

Phenol, Molecular Biology Grade

Synonym(s):

Phenol, C₆H₆O, Hydroxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OH
CAS Number:
Molecular Weight:
94.11
Beilstein:
969616
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.25
form:
crystalline solid
Assay:
≥99% (GC)

grade

Molecular Biology

Quality Level

vapor density

3.24 (vs air)

vapor pressure

0.09 psi ( 55 °C)
0.36 mmHg ( 20 °C)

Assay

≥99% (GC)

form

crystalline solid

autoignition temp.

1319 °F

expl. lim.

8.6 %

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

bp

182 °C (lit.)

mp

40-42 °C (lit.)

solubility

water: soluble

density

1.071 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.0002%
Mg: ≤0.0003%
heavy metals: ≤0.0008%

foreign activity

DNase, RNase, none detected

storage temp.

−20°C

SMILES string

Oc1ccccc1

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H

InChI key

ISWSIDIOOBJBQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Useful for the purification of nucleic acids.

Warning

Toxicity: Toxic (F)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

Target Organs

Nervous system,Kidney,Liver,Skin

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service