Skip to Content
Merck
All Photos(1)

Key Documents

5161

Millipore

Penicillin G, Potassium Salt

Antibiotic effective against Gram-positive bacteria. Blocks cell wall formation in growing cells, resulting in accumulation of uracil nucleotides of muramic acid peptides.

Synonym(s):

Penicillin G, Potassium Salt, Benzylpenicillin, K

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17N2O4S · K
CAS Number:
Molecular Weight:
372.48
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 100 units/mL (suggested concentration)

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C17H19ClN2S.C16H18N2O4S.K/c1-19(2)10-5-11-20-14-6-3-4-7-16(14)21-17-9-8-13(18)12-15(17)20;1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-4,6-9,12H,5,10-11H2,1-2H3;3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;;+1/p-1/t;11-,12+,14-;/m.1./s1

InChI key

HTQQVOYPSGNVPA-HBRCYENSSA-M

General description

Antibiotic effective against Gram-positive bacteria. Blocks cell wall formation in growing cells, resulting in accumulation of uracil nucleotides of muramic acid peptides. Acts at the final stage of cell wall synthesis and prevents cross-linking of peptidoglycan strands.
Antibiotic effective against gram-positive and gram-negative bacteria. Blocks cell wall formation in growing cells, resulting in the accumulation of uracil nucleotides of muramic acid peptides. Acts at the final stage of cell wall synthesis and prevents cross-linking of peptidoglycan strands.
Note: 1 MU = 1,000,000 units.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is equivalent to 0.6 µg of penicillin G.

Reconstitution

Following reconstitution, refrigerate (4°C). Stock solutions are stable for up to 1 week at 4°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service