Skip to Content
Merck
All Photos(1)

Key Documents

5.09453

Sigma-Aldrich

JHDM Inhibitor VII, JIB-04

Synonym(s):

JHDM Inhibitor VII, JIB-04, 5-Chloro-2-((2 E)-2-(phenyl(2-pyridinyl)methylene)hydrazino)pyridine, Histone Lysine Demethylase Inhibitor XIII, NSC693627, JIB04

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClN4
CAS Number:
Molecular Weight:
308.76
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

InChI

1S/C17H13ClN4/c18-14-9-10-16(20-12-14)21-22-17(13-6-2-1-3-7-13)15-8-4-5-11-19-15/h1-12H,(H,20,21)/b22-17+

InChI key

YHHFKWKMXWRVTJ-OQKWZONESA-N

General description

The (E)-isomer of a cell-permeable pyridine hydrazone compound that acts as a selective inhibitor against Jumonji family histone demethylases (IC50 = 230, 290-320, 340, 435, 445, 855, and 1,100 nM, respectively, against JARID1A/KDM5A, JMJD2D/KDM4D, JMJD2E/KDM4E, JMJD2B/KDM4B, JMJD2A/KMD4A, JMJD3/KDM6B, and JMJD2C/KDM4C, respectively), presumably via catalytic site iron interaction, displaying little or no potency toward 5mC DNA hydroxylase TET1, aurora kinases, HDACs, HIF prolyl hydroxylase PHD2, histone methyltransferases, protein arginine methyltransferases, sirtuins, or the non-jumonji family histone demethylase LSD1/KDM1A. Shown to exhibit anticancer activity against H358 and A549 lunger cancer cells both in cultures in vitro (IC50 = 100 and 250 nM, respectively, in 4 d by MTS assays) and in mice in vivo (via i.p. at 100 mg/kg and p.o. at 55 mg/kg, respectively; 2-3 times per week).
The E-isomer of a cell-permeable pyridine hydrazone compound that acts as a selective inhibitor against Jumonji family histone demethylases (IC50 = 230, 290-320, 340, 435, 445, 855, and 1,100 nM, respectively, against JARID1A/KDM5A, JMJD2D/KDM4D, JMJD2E/KDM4E, JMJD2B/KDM4B, JMJD2A/KMD4A, JMJD3/KDM6B, and JMJD2C/KDM4C, respectively), presumably via catalytic site iron interaction, displaying little toward 5mC DNA hydroxylase TET1, aurora kinases, HDACs, HIF prolyl hydroxylase PHD2, histone methyltransferases, protein arginine methyltransferases, sirtuins, or the non-jumonji family histone demethylase LSD1/KDM1A. Shown to exhibit anticancer activity against H358 and A549 lunger cancer cells both in cultures in vitro (IC50 in 4 d = 100 and 250 nM, respectively) and in mice in vivo (via i.p. at 100 mg/kg and p.o. at 55 mg/kg, respectively; 2-3 times per week).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Jumonji family of histone demethylases
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Wang, L., et al. 2013. Nat. Commun.4, 2035.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service