5.05760
HDAC6 Inhibitor II, BRD9757
Synonym(s):
HDAC6 Inhibitor II, BRD9757, N-Hydroxycyclopent-1-enecarboxamide, HDAC Inhibitor XVI, Histone Deacetylase Inhibitor XVI
About This Item
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
pale orange
solubility
DMSO: 100 mg/mL
storage temp.
2-8°C
General description
Biochem/physiol Actions
HDAC6
Packaging
Warning
Reconstitution
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service