Skip to Content
Merck
All Photos(1)

Key Documents

5.04480

Sigma-Aldrich

Conantokin G

Synonym(s):

Conantokin G, Gly-Glu-γ-CarboxyGlu-γ-CarboxyGlu-Leu-Gln-γ-CarboxyGlu-Asn-Gln-γ-CarboxyGlu-Leu-Ile-Arg-γ-CarboxyGlu-Lys-Ser-Asn-NH2

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

Assay

≥95% (HPLC)

Quality Level

form

powder

potency

0.8 μM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 1 mg/mL

storage temp.

−20°C

InChI

1S/C88H138N26O44/c1-7-34(6)61(77(138)103-41(12-10-20-98-88(96)97)63(124)107-48(23-35(78(139)140)79(141)142)69(130)100-40(11-8-9-19-89)64(125)113-54(31-115)76(137)104-45(62(95)123)28-57(93)118)114-75(136)47(22-33(4)5)106-70(131)49(24-36(80(143)144)81(145)146)109-67(128)44(14-17-56(92)117)102-74(135)53(29-58(94)119)112-73(134)51(26-38(84(151)152)85(153)154)110-66(127)43(13-16-55(91)116)101-68(129)46(21-32(2)3)105-71(132)52(27-39(86(155)156)87(157)158)111-72(133)50(25-37(82(147)148)83(149)150)108-65(126)42(15-18-60(121)122)99-59(120)30-90/h32-54,61,115H,7-31,89-90H2,1-6H3,(H2,91,116)(H2,92,117)(H2,93,118)(H2,94,119)(H2,95,123)(H,99,120)(H,100,130)(H,101,129)(H,102,135)(H,103,138)(H,104,137)(H,105,132)(H,106,131)(H,107,124)(H,108,126)(H,109,128)(H,110,127)(H,111,133)(H,112,134)(H,113,125)(H,114,136)(H,121,122)(H,139,140)(H,141,142)(H,143,144)(H,145,146)(H,147,148)(H,149,150)(H,151,152)(H,153,154)(H,155,156)(H,157,158)(H4,96,97,98)

InChI key

HTBKFGWATIYCSF-UHFFFAOYSA-N

General description

The most selective and potent non-competitive antagonist among the members of the conantokin peptide family for NR2B-containing NMDAR channels (Ki = 0.8 µM with close to 100% inhibition of NR1/NR2B mediated current). Widely used for studies related to NMDAR neurotransmissions in both peripheral and center nervous systems.

Biochem/physiol Actions

Primary Target
NMDA receptors

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

Gly-Glu-γ-CarboxyGlu-γ-CarboxyGlu-Leu-Gln-γ-CarboxyGlu-Asn-Gln-γ-CarboxyGlu-Leu-Ile-Arg-γ-CarboxyGlu-Lys-Ser-Asn-NH₂

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Sheng, Z., et al. 2007. Neuropharmacology.53, 145.
Sheng, Z., et al. 2009. Neuropharmacology.57, 127.
Zhou, L., et al. 1996. J. Neurochemistry.66, 620.
Hammerland, L., et al. 1992. Eur. J. Pharmacol.226, 239.

Mena, E., et al. 1990. Neurosci Lett.118, 241.
Donevan, D., et al. 2000. Mol Pharmacol.58, 614.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service