Skip to Content
Merck
All Photos(2)

Documents

481417

Sigma-Aldrich

CRAC Channel Inhibitor IV, EVP4593

The NF-κB Activation Inhibitor VIII, EVP4593 controls the biological activity of NF-κB. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

CRAC Channel Inhibitor IV, EVP4593, N4-(4-phenoxyphenethyl)quinazoline-4,6-diamine, SOC Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N4O
Molecular Weight:
356.42
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL, clear, yellow

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable quinazoline derivative that blocks PMA/PHA-induced NF-κB activation in Jurkat cells (IC50 = 11 nM), but has no effect on PKC kinase, and exhibits an anti-inflammatory effect on carrageenin-induced paw edema in rats (1 mg/kg, i.p.). In addition, it is shown to inhibit the store-operated calcium entry (SOC) pathway possibly by targeting TRPC1 containing heteromeric channels in Huntington Disease (HD) neurons (SK-N-SH cells transfected with mutant Huntingtin Htt-138Q) at 300 nM. It displays neuroprotective effects in YAC128 MSN mice neurons from glutamate toxicity dose-dependently from 30 nM to 3 µM, which is consistent with TRPC1 knockdown effects, and attenuates the progressive decline in the climbing speed of the HD Drosophila, dose-dependently from 100 µM to 400 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, J., et al. 2011. Chem Biol.18, 777.
Tobe, M., et al. 2003. Bioorg. Med. Chem.11, 383.
Tobe, M., et al. 2003. Bioorg. Med. Chem.11, 3869.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dmitriy A Grekhnev et al.
International journal of molecular sciences, 23(24) (2022-12-24)
Quinazoline derivatives have various pharmacological activities and are widely used in clinical practice. Here, we reviewed the proposed mechanisms of the physiological activity of the quinazoline derivative EVP4593 and perspectives for its clinical implication. We summarized the accumulated data about

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service