Skip to Content
Merck
All Photos(1)

Key Documents

420311

Millipore

Kanamycin Sulfate, Streptomyces kanamyceticus

An aminoglycoside antibiotic effective against Gram-positive and Gram-negative organisms.

Synonym(s):

Kanamycin Sulfate, Streptomyces kanamyceticus, Kamycin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N4O11 · xH2SO4
CAS Number:
Molecular Weight:
484.50 (free base basis)
UNSPSC Code:
51281654
Pricing and availability is not currently available.

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

10-30°C

InChI

1S/C18H36N4O11.H2O4S/c19-2-6-10(25)12(27)13(28)18(30-6)33-16-5(21)1-4(20)15(14(16)29)32-17-11(26)8(22)9(24)7(3-23)31-17;1-5(2,3)4/h4-18,23-29H,1-3,19-22H2;(H2,1,2,3,4)/t4-,5+,6-,7-,8+,9-,10-,11-,12+,13-,14-,15+,16-,17-,18-;/m1./s1

InChI key

OOYGSFOGFJDDHP-KMCOLRRFSA-N

General description

Contains more than 98% kanamycin A. An aminoglycoside antibiotic affective against gram-positive and gram-negative organisms. Inhibitor of protein biosynthesis that acts on the 70S ribosome, causing misreading of the genetic code. May cause renal damage and ototoxicity.
Potency: ≥750 µg/mg

Biochem/physiol Actions

Primary Target
protein biosynthesis

Reconstitution

Following reconstitution, sterilize by filtration through a 0.22 µm pore-size filter, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Lech, K., and Brent, R. 1995. in Current Protocols in Mol. Biol. Vol. 1 (Ausubel, F.M., et al., eds.) John Wiley & Sons, New York.
Kahn, R.W., et al. 1993. Proc. Natl. Acad. Sci. USA90, 9295.
Cardon, G.H., et al. 1993. Plant J.3, 773.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition. Cold Spring Harbor, New York. p.1.6, A.6.
Korzybyski, T., et al. 1978. In Antibiotics, Origin, Nature, and Properties1, 690.
Stiffler, P.W., et al. 1974. Antimicrob. Agents Chemother.6, 516.
Umezawa, S., et al. 1968. J. Antibiot.21, 367.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service