Skip to Content
Merck
All Photos(1)

Key Documents

401459

Sigma-Aldrich

Mouse Anti-Human IgG₁, Fc Fragment Specific Alkaline Phosphatase Conjugate

liquid, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

secondary antibodies

clone

monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

Protein A purified mouse monoclonal antibody conjugated to alkaline phosphatase. Recognizes human IgG1, Fc fragment.
This Mouse Anti-Human IgG₁, Fc Fragment Specific Alk-Phos Conjugate is validated for use in ELISA, Enzyme Immunoassay, Immunoelectrophoresis for the detection of Human IgG₁.

Application

ELISA (see comments)

Enzyme Immunoassay (≥1:5000)

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 50 mM Tris, 1 mM MgCl₂, 50% glycerol, pH 8.0.

Other Notes

Monospecific for human IgG1 Fc fragment as determined by immunoelectrophoresis and direct ELISA against normal human serum and normal human IgG. p-Nitrophenylphosphate, 1 mg/ml in 10% diethanolamine, pH 9.8, 1 mM MgCl2 at 25°C, was used as substrate for testing immunoenzyme reactivity. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yu et al.
Frontiers in cellular and infection microbiology, 14, 1342621-1342621 (2024-02-19)
Murine research has revealed a significant role for antibody responses in protection against Chlamydia reinfection. To explore potential humoral immune markers of protection elicited by Chlamydia trachomatis (CT) antigens in humans in the context of presumed clinical correlates of protection
Shota Kobayashi et al.
Journal of neuroimmunology, 257(1-2), 102-106 (2013-01-15)
Autoantibody against nicotinic acetylcholine receptor (nAChR) α3 subunit has been implicated in the pathogenesis of paraneoplastic neurological syndrome. To examine the effect of anti-α3 subunit autoantibody on cell-surface nAChRs, we established human embryonic kidney 293 cells stably co-expressing α3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service