Skip to Content
Merck
All Photos(1)

Documents

375230

Sigma-Aldrich

HGF, Mouse, Recombinant

HGF, Mouse, Recombinant Recombinant, consists of disulfide-linked heterodimeric α (Gln33 to Arg495) and β (Val496 to Leu728) chains expressed in murine myeloma NS0 cell line.

Synonym(s):

Hepatopoietin A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

Quality Level

Assay

≥95% (as seen by a 60 kDa (α chain) and a 33 kDa (β chain) bands under reducing conditions., SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse hepatocyte growth factor (HGF) consisting of disulfide-linked heterodimeric α (Gln33 to Arg495) and β (Val496 to Leu728) chains expressed in murine myeloma NS0 cell line.

Biochem/physiol Actions

Cell Proliferation:

ED₅₀ = 10-30 ng/ml as measured in a cell proliferation assay using IMCD-3 mouse epithelial cells.



Functional ELISA

KD<0.2 nM as measured by binding to C-Met (HGF Receptor).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Reconstitute to 10 µg/ml in sterile PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Corso, S., et al. 2005. Trends Mol. Med.11, 284.
Karihaloo, A., et al. 2005. Nephron Exp. Nephrol.100, e40.
Urbanek, K., et al. 2005. Circ. Res.97, 663.
Hammond, D.E., et al. 2004. Curr. Top. Microbiol. Immunol.286, 21.
Rosario, M. and W. Birchmeier. 2004. Dev. Cell7, 3.
Gheradi, E., et al. 2003. Proc. Natl. Acad. Sci.100, 12039.
Chiu, S.J., et al. 2002. J. Biomed. Sci.9, 261.
Tanaka, Y., et al. 2002. Biochem. Biophys. Res. Commun.299, 472.
Maeshima, A., et al. 2000. Kid. Int.58, 1511.
Lesk, A.M. and W.D. Fordham. 1996. J. Mol. Biol.258, 501.
Saelman, E.U.M., et al. 1995. J. Cell Sci.108, 3531.
Mineo, R., et al. 1994. Endocrinology145, 4355.
Mizuno, K., et al. 1994. J. Biol. Chem.269, 1131.
Sasaki, M., et al. 1994. Biochem. Biophys. Res. Commun.199, 772.
Montesano, R., et al. 1991. Cell67, 901.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service