Skip to Content
Merck
All Photos(1)

Key Documents

344911

Sigma-Aldrich

Fura-2 LR/AM

Acetoxymethyl (AM) derivative of FURA-PE3 with spectral properties similar to the parent compound.

Synonym(s):

Fura-2 LR/AM, Fura-2 Leakage Resistant (AM)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H63N5O29
CAS Number:
Molecular Weight:
1258.10
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

gum

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 10 mg/mL

fluorescence

λex 368 nm (low)
λex 374 nm (high)
λem 518 nm (low)
λem 524 nm (high)

shipped in

ambient

storage temp.

−20°C

SMILES string

N1(CCN(CC1)C(=O)CCc2cc(c(cc2)N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)OCCOc3cc4c([o]c(c4)c5ncc([o]5)C(=O)OCOC(=O)C)cc3N(CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C)CC(=O)OCOC(=O)C

InChI

1S/C55H63N5O29/c1-33(61)76-27-82-49(68)22-57-11-13-58(14-12-57)48(67)10-8-39-7-9-41(59(23-50(69)83-28-77-34(2)62)24-51(70)84-29-78-35(3)63)44(17-39)74-15-16-75-45-18-40-19-46(54-56-21-47(89-54)55(73)87-32-81-38(6)66)88-43(40)20-42(45)60(25-52(71)85-30-79-36(4)64)26-53(72)86-31-80-37(5)65/h7,9,17-21H,8,10-16,22-32H2,1-6H3

InChI key

CGAZNAYDEAJZHQ-UHFFFAOYSA-N

General description

Acetoxymethyl (AM) derivative of FURA-PE3 with spectral properties similar to the parent compound. Can be loaded into cells in a manner similar to FURA 2/AM. Hydrolysis by non-specific esterases traps the indicator in the cytosol. Compared to FURA-2, it is retained longer in the cell. Note: 1 set = 10 x 50 µg.
An acetoxymethyl (AM) derivative of FURA-PE3. with spectral properties similar to the parent compound. Can be loaded into cells in a manner similar to FURA 2/AM (Cat. No. 344905). Hydrolysis by non-specific esterases traps the indicator in the cytosol. Spectral properties in the presence of low calcium concentrations (ex. 364 nm, em. 508 nm) differ from those during high calcium conditions (ex. 335 nm, em. 495 nm). Note: 1 set = 10 x 50 µg.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Calcium Probes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Abe, F., et al. 1995. Br. J. Pharmacol.116, 3000.
Vorndran, C., et al. 1995. Biophys. J.69, 2114.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service