Skip to Content
Merck
All Photos(1)

Key Documents

344273-M

Sigma-Aldrich

Forskolin, 7-Deacetyl-7-[O-(N-methylpiperazino)-γ-butyryl]-, Dihydrochloride

≥92% (HPLC), solid, adenylate cyclase activator, Calbiochem®

Synonym(s):

Forskolin, 7-Deacetyl-7-[O-(N-methylpiperazino)-γ-butyryl]-, Dihydrochloride, L-858051, 8,13-Epoxy-7β-(N-methylpiperazino-γ-butyryloxy)-1α,6β,9α-trihydroxy-labd-14-en-11-one, 2HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H48N2O7 · 2HCl
CAS Number:
Molecular Weight:
609.62
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Forskolin, 7-Deacetyl-7-[O-(N-methylpiperazino)-γ-butyryl]-, Dihydrochloride, Chemically-modified forskolin with greater stability and water solubility than the parent compound.

Quality Level

Assay

≥92% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 12 mg/mL
methanol: soluble

storage temp.

10-30°C

SMILES string

Cl.Cl.N4(CCN(CC4)C)CCCC(=O)O[C@@H]1[C@]2(O[C@@](CC(=O)[C@@]2([C@@]3([C@@H]([C@@H]1O)C(CC[C@@H]3O)(C)C)C)O)(C)C=C)C

InChI key

OAJWVIUBCCGPBN-BAPSXVLWSA-N

General description

A chemically modified forskolin with greater stability and water solubility. Activates adenylate cyclase (EC50 = 13 µM).
Chemically-modified forskolin with greater stability and water solubility than the parent compound. Activates adenylate cyclase (EC50 = 13 µM). Inhibits 5HT-stimulated inositol phosphate accumulation in smooth muscle cells.

Biochem/physiol Actions

EC50 = 13 µM in activating adenylate cyclase
Primary Target
Adenylate cyclase

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 4 months at 4°C.

Other Notes

Yang, C.M., et al. 1999. Cell Signal. 11, 697.
Hartzell, H.C., and Budnitz, D. 1992. Mol. Pharmacol.41, 880.
Chidiac, P., et al. 1991. J. Mol. Cell. Cardiol.23, 1255.
Laurenza, A., et al. 1987. Mol. Pharmacol.32, 133.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service