Skip to Content
Merck
All Photos(1)

Key Documents

34321-M

Sigma-Aldrich

FITC, Isomer I

Synonym(s):

FITC, Isomer I, Fluorescein Isothiocyanate, Isomer I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H11NO5S
CAS Number:
Molecular Weight:
389.38
MDL number:
UNSPSC Code:
41116133
Pricing and availability is not currently available.

Assay

≥95% (TLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

orange-red

solubility

DMSO: 5 mg/mL
DMF: soluble
aqueous buffer, pH ≥ 9.0: soluble

storage temp.

2-8°C

SMILES string

S=C=Nc1cc2c(cc1)C3(OC2=O)c4c(cc(cc4)O)Oc5c3ccc(c5)O

InChI

1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H

InChI key

MHMNJMPURVTYEJ-UHFFFAOYSA-N

General description

A fluorescent labeling agent used to label antibodies, lectins, toxins, various proteins, and oligopeptides with fluorescein. FITC-antibody conjugates are useful in many applications, including flow cytometry and immunohistochemistry (excitation maximum = 492 nm; emission maximum = 520 nm). λ max: 492 ± 3 nm (pH 9). Fluorescence EMmax: 520 ± 3 nm (pH 9).
Popular derivative of fluorescein. Can be used to label antibodies and other proteins.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cherry, N.B., et al. 1965. Stain Technol. 44, 179.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is there a SDS available for this product?

    1 answer
    1. Please access the Material Safety Data Sheet in the link below:
      https://www.emdmillipore.com/product/msds/EMD_BIO-34321?Origin=SERP

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service